Learn More
Thermo Scientific™ Thermo Scientifi™ Chiller External Pressure Reducers
Shop All Fisherbrand ProductsDescription
External pressure reducers for ThermoChill Recirculating chillers
- 1/2 in. external pressure reducer
- Adjustable to safely reduce or limit system pressure and flow
Specifications
Specifications
| For Use With (Equipment) | ThermoChill III |
| Type | External Pressure Reducer (EPR) |
| Size | 1/2 in. |