Includes
Batteries, with lighted models.
Fisher Scientific™ Hands-Free Fold-Up Magnifiers all fold twice for compact storage. Allow two-handed examination of objects while yielding a sharp, crisp and distinct image.
- Bottom panel features white grid markings in both inches and millimeters to assist in measuring
- All-glass optics ensure perfect focus with zero distortion
- Seven sizes are available, including three featuring brilliant battery-powered illumination
Specifications
Specifications
| For Use With (Application) | For use in quality control, labs, food, electronics, manufacturing and universities |
| Diameter (Englis) Lens | 2 in. |
| Diameter (Metric) Lens | 5.08 cm |
| Magnification | 3.5x |
| Material | Glass |
| Product Type | Magnifier (Hands-free, Fold-up) |