Learn More
Assorted pH Buffer Pouches, Fisherbrand™
Product Code. 17927242
Change view
Click to view available options
Quantity:
20 pouches
Unit Size:
1 set
Product Code. Quantity unitSize
17927242 20 pouches 1 set
1 options
Product Code. 17927242
Brand: Fisherbrand 3565304
Economical, accurate, convenient
  • ±0.01pH accuracy at 25°C (77°F)
  • Pack of 20 pouches: five pouches each of pH 4, 7, and 10 buffers and rinse solution
Certifications
All solutions are certified traceable to NIST™ standards.
Specifications
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 4; 10; 7
Physical Form Liquid
Quantity 20 pouches
Solution Type Buffer