Description
- For monitoring temperature sensitive samples volumes
- Can be used to monitor volumes from 100 - 1000mL
- 6 in. long Temperature monitoring probe (Stainless Steel)
Specifications
Specifications
| Product Type | Temperature Probe |
| Length (English) | 6 in. |
| Length (Metric) | 15.24 cm |
| For Use With (Application) | Ideal for monitoring temperature of samples 100mL or larger |
| For Use With (Equipment) | FB 705 Sonic Dismembrator™ |