Learn More
Fisherbrand™ Hand Tally Counter
Shop All Fisherbrand Products
Click to view available options
Displayed Text:
9999
Unit Size:
Each
Beskrivelse
- Easy-to-use, four-digit unit records up to 9999
- Numbers instantly and automatically return to zero by twisting the reset knob
- Large white numerals appear on a black background
- Protective window seals out dust, dirt and humidity
- Rugged, heavy-duty chrome-finished case has a hinged finger loop for a sure grip
- Finger pressure on the button produces an audible click that signals the count
- Unique engineering provides a smooth, tactile feel with every count
- Dimensions: 1.75 dia. × 1.13 in.W (4.4 × 2.9cm)
- Weight: 3 oz.
Tekniske data
Tekniske data
| Product Type | Hand Tally Counter |
| Displayed Text | 9999 |
| Quantity | Each |
| Diameter (English) | 1.75 in. |
| Diameter (Metric) | 4.45cm |
| Width (English) | 1.13 in. |
| Width (Metric) | 2.9cm |
| For Use With (Application) | Ideal for lab applications |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion