Learn More
Fisher BioReagents™ exACTGene™ DNA Ladders
Click to view available options
No. of Bands:
8
10
11
12
13
14
15
16
19
For Use With (Application):
Clone Identification
Fast run times, small size DNA
General purpose, extra large size
General purpose, large digested DNA
General purpose, small size DNA
General purpose, wide size range
Large size PCR or cloning
PCR size confirmation
Quick check of PCR or Enzyme digestion
Small and large cloning
Unit Size:
Each
Description
Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze
- Specially engineered to provide blunt-end DNA of precise length for better band quality than classic markers
- Ideal for size assessment and quantitative determination of DNA mass
- Contain loading dye to reduce pipetting steps and save time
- Have two or three high intensity reference bands for better sizing
- Provided in sufficient quantity to load 100 lanes
Specifications
Specifications
| Color | Colorless |
| Product Type | DNA Molecular Weight Marker/Ladder |
| Form | Liquid |
| Formulation | Premixed with loading dye |
| For Use With (Application) | General purpose, wide size range |
| No. of Bands | 19 |
| Quantity | 100 Lanes |
| Storage Requirements | Stable at room temperature for two years. |
| Sufficient For | 100 Lanes |