Learn More
Fisher BioReagents™ exACTGene™ DNA Ladders
Product Code. 10489883
Click to view available options
No. of Bands:
8
10
11
12
13
14
15
16
19
For Use With (Application):
Clone Identification
Fast run times, small size DNA
General purpose, extra large size
General purpose, large digested DNA
General purpose, small size DNA
General purpose, wide size range
Large size PCR or cloning
PCR size confirmation
Quick check of PCR or Enzyme digestion
Small and large cloning
Unit Size:
Each
Product Code. 10489883
Brand: Fisher BioReagents™ BP2578100
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze
  • Specially engineered to provide blunt-end DNA of precise length for better band quality than classic markers
  • Ideal for size assessment and quantitative determination of DNA mass
  • Contain loading dye to reduce pipetting steps and save time
  • Have two or three high intensity reference bands for better sizing
  • Provided in sufficient quantity to load 100 lanes
TRUSTED_SUSTAINABILITY
Specifications
Color Colorless
Product Type DNA Molecular Weight Marker/Ladder
Form Liquid
Formulation Premixed with loading dye
For Use With (Application) General purpose, large digested DNA
No. of Bands 13
Quantity 100 Lanes
Storage Requirements Stable at room temperature for two years.
Sufficient For 100 Lanes