missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIS1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA141082
This item is not returnable.
View return policy
Description
This antibody is predicted to react with rat, chimp, and canine based on sequence homology. Suggested positive control: antigen standard for FIS1 (transient overexpression lysate), 3T3 or Hela whole cell lysate.
Fis1, a 17KDa integral membrane protein, is a novel member of mitochondrial fission machinery. It consists of a central leucine-zipper domain, a coiled-coil region, a tetratricopeptide repeat domain and a C-terminal transmembrane domain. It is a human orthologue of Fis1p (S.cerevisiae) and is localized in the outer mitochondrial membrane. Fis1 acts as a pro-apoptotic factor and plays a vital role in the regulation of mitochondrial fission. It interacts with DLP1 and Mdv1p and facilitates Dnm1p recruitment to the functional fission complex, thereby inducing apoptosis. Role of Fis1 has also been implicated in growth and division of peroxisomes. Loss of Fis1 inhibits apoptosis and results in elongation of mammalian mitochondria.
Specifications
| FIS1 | |
| Polyclonal | |
| Unconjugated | |
| FIS1 | |
| 2010003O14Rik; CGI-135; Fis1; fis1 {ECO:0000250; FIS1 homolog; fission 1 (mitochondrial outer membrane) homolog; fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae); fission 1 (mitochondrial outer membrane) homolog (yeast); fission, mitochondrial 1; H_NH0132A01.6; hFis1; Mitochondrial fission 1 protein; mitochondrial fission molecule; rFis1; tetratricopeptide repeat domain 11; tetratricopeptide repeat protein 11; TPR repeat protein 11; TTC11; UniProtKB:Q9Y3D6} | |
| Rabbit | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Conc. Not Determined | |
| whole serum diluted in PBS with 0.05% BSA and 0.05% sodium azide | |
| P84817, Q9CQ92, Q9Y3D6 | |
| FIS1 | |
| Synthetic peptide corresponding to residues 11-25 of human Fis1. | |
| RUO | |
| 288584, 51024, 66437 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction