missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Firefly luciferase Monoclonal Antibody (Luci 21 1-107)
GREENER_CHOICE

Product Code. 11574412
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
11574412 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 11574412

Brand: Invitrogen™ MA116880

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Suggested positive control: purified Luciferase protein or Luciferase expressed in Drosophila.

Luciferase produces green light with the wavelength of 562 nm. Luciferase catalyzes the reaction: D-firefly luciferin + O2 + ATP = firefly oxyluciferin + CO2 + AMP + diphosphate + light.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Firefly luciferase
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone Luci 21 1-107
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias Firefly Luciferase; Genji firefly; Green Luciferase; Japanese firefly; luciferin 4-monooxygenase; Photinus pyralis; photinus pyralis luciferase
Host Species Mouse
Immunogen Luciferase protein from Photinus pyralis (North American firefly).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse, Luciferase
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.