Learn More
Thermo Scientific™ Finnpipette™ Multistepper Pipette
Product Code. 10591151
Click to view available options
Volume (Metric):
1250 μL
50 to 250 μL
Unit Size:
Each
Product Code. 10591151
Brand: Thermo Scientific™ 2206590
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Reduce the risk of error and increase productivity in immunoassay applications with this multistepper pipette, designed especially for work with microplates and featuring simultaneous 8-channel dispensing.
Reduce the risk of error and increase productivity in immunoassay applications with the Thermo Scientific™ Finnpipette™ Multistepper Pipette. This lightweight, easy-to-use repeater pipette provides simultaneous dispensing in eight channels. Designed especially for work with microplates, the Finnpipette Multistepper helps speed up routine laboratory procedures and is easy to use — simply attach, fill, and prime the module, then dial the selected volume.
Design
  • Universal handle also fits the seven single-channel Thermo Scientific™ Finntip™ Steppers
  • Lightweight construction minimizes hand fatigue during repetitive dispensing
  • Separate tip ejector for safe and effortless tip disposal
  • Wide finger rest minimizes operator hand strain
  • Module rotates a full 360°, optimizing efficiency for both right or left-handed users
Compatible with:
Accepts Finntip Multistepper 1500μL tips. They are extra long, with more air space between the liquid and tip cone to prevent contamination.
TRUSTED_SUSTAINABILITY
Specifications
Description 8-Channel Module only
For Use With Microplate
Number of Channels 8
Type Multistepper Pipette
Volume (Metric) 1250 μL