Learn More
Thermo Scientific™ Finnpipette™ F1 Multichannel Pipettes
Product Code. 15591925
Change view
Click to view available options
Volume (Metric):
1 to 10 μL
10 to 100 μL
30 to 300 μL
5 to 50 μL
Number of Channels:
8
12
16
Color:
Orange
Pink
Purple
Turquoise
Yellow
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Number of Channels Color Unit Size
15591925 30 to 300 μL 8 Orange Each
15561925 1 to 10 μL 8 Pink Each
15571925 5 to 50 μL 8 Yellow Each
15581925 10 to 100 μL 8 Yellow Each
15501935 1 to 10 μL 12 Pink Each
15511935 5 to 50 μL 12 Yellow Each
15521935 10 to 100 μL 12 Yellow Each
15531935 30 to 300 μL 12 Orange Each
15541935 1 to 10 μL 16 Purple Each
15551935 5 to 50 μL 16 Turquoise Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15591925 Supplier Thermo Scientific™ Supplier No. 4661030N
  • ONLINE EXCLUSIVE PRICE
    12996.59 NOK
    11570.24 NOK / Pack of 1
    Save 1426.35 NOK
    10% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request
Get optimal performance and ergonomic comfort from these F1 Multichannel Pipettes. Available in 8-, 12- or 16-channel models; choose from a vast selection of Finntip pipette tips for optimal results.
Get optimal performance and ergonomics from the Finnpipette™ F1 system, which delivers performance with precision, all in a lightweight design that maximizes user comfort and reduces strain. It's the ideal system for applications using expensive reagents and valuable samples.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F1: Everything you want in your everyday, go-to pipette
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI
Silent, Effortless Volume Adjustment
Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.
Secure Volume Lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50 μl models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
CE marked
The Finnpipette F1 Pipette together with compatible Finntip tips is CE marked in accordance with IVD Directive in Europe.
5 Year Warranty
With convenient online registration, and may vary by country.
Learn more about the complete Finnpipette family >
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips Finntip 300, Filter 300, Flex 300, Flex Filter 300
Description Finnpipette F1
Color Orange
Number of Channels 8
Volume (Metric) 30 to 300 μL
Increments 1μL
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Datenschutzrichtlinie.