Learn More
Thermo Scientific™ Finger Tite Ferrules
Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric):
0.4 mm
0.5 mm
Unit Size:
Pack of 10
Description
Thermo Scientific™ Finger Tite Ferrules offer easier fit, reliabililty and leak-free connections.
Thermo Scientific Finger Tite ferrules are the next generation of the SilTite™ connection system.
Advantages over conventional graphite/polymer ferrules:
- Perfect metal-to-metal seal eliminates contamination and gives lower air/water background
- No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures
- No tools required
Specifications
Specifications
| Type | Ferrule |
| Description | 0.4 mm ferrule |
| Diameter (Metric) | 0.4 mm |
| For Use With (Equipment) | 0.1 to 0.25 mm I.D. GC Columns |