Få mere at vide
Thermo Scientific™ Finger Tite Connectors
Shop alle Thermo Scientific produkterBeskrivelse
Ensure reliable, leak-free connections for optimal GC performance using Thermo Scientific™ Finger Tite Connectors. The perfect metal-to-metal seal eliminates contamination and produces lower air/water background noise compared to conventional graphite/polymer ferrules. They are easy to handle even with a hot injector/detector and do not require the use of tools. Unlike other ferrules, they do not require retightening after installation.
- Thermo Scientific™ and Agilent™ GC instruments
- Connectors for SSL and MS systems
Tekniske data
Tekniske data
| Type | Split/Splitless and MS Starter Kit |
| Description | MS Starter Kit |
| For Use With (Equipment) | TRACE GC Ultra and FOCUS GC Instruments |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.