Learn More
Ekastu Safety™ Mandil Filtering Facepiece FFP1/V with Cool Down Exhalation Valve
Product Code. 10299451
Product Code. 10299451
Brand: Ekastu Safety™ 411381
Protects against fine inert particles up to 4.5 times the NPF value. EKASTU Safety™ Mandil Filtering Facepiece FFP1/V with Cool Down Exhalation Valve has a five-year shelf life in original packaging.
  • With Cool Down exhalation valve
  • With colour-coded nosepiece and exhalation valve to immediate identification for the protection class protection against fine inert particles up to 4.5 times the NPF value
  • Filter class DIN EN 149:2001 FFP1 NR
  • In original packaging: 5 years shelf life
  • 12 pieces per pack
Specifications
Color Yellow
Resistant To 160 l/min, constant flow max. 3.0 mbar