Learn More
Epredia™ Fine Pore Cassettes
Product Code. 16334096 Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Product Code. Color unitSize
16334096 Orange Pack of 1000
16364086 Beige Pack of 1000
16374086 Blue Pack of 1000
16384086 Green Pack of 1000
16394086 Gray Pack of 1000
16304096 Lavender Pack of 1000
16314096 Light Green Pack of 1000
16324096 Lilac Pack of 1000
16344096 Pink Pack of 1000
16354096 Tan Pack of 1000
16364096 White Pack of 1000
16374096 Yellow Pack of 1000
12 options
Product Code. 16334096
Brand: Epredia™ B851735OR
For very small biopsy tissue samples, cassettes have a front-edge lid hinge with attached lids that are available in 12 different color varieties
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Fine Pore
Closure Included Yes
Packaging Type Loose Packed
Quantity 1000/Cs.
Closure Type Attached Lid, Front Hinge
PrintMate Compatible No
Color Orange
Product Type Cassette