Description
The Findenser™ consists of a glass column and a finned aluminum housing jacket. A small amount of encapsulated thermal fluid located between the glass column and aluminum jacket transfers heat from the glass to the finned aluminum jacket. The heat generated in the process is then dissipated to the outside via the innovative structure of the aluminum fins. When used in the laboratory fume hood, the Findenser™ offers a cooling capacity that is in no way inferior to water-cooled systems.
- For syntheses under reflux
- Large surface of aluminum fins guarantees excellent heat transfer
- Use with solvent volumes up to 125 mL
- Can be secured with standard laboratory clamps or with Findenser™ Attachment Kit (sold separately)
Specifications
Specifications
| For Use With (Application) | Condensing Water |
| Length (Metric) | 275 mm |
| Material | Glass, Aluminum |
| For Use With (Equipment) | Laboratory Glassware |
| Length (English) | 10.8 in. |
| Standard Taper Joint | NS 19/19 |
| Joint Type | Cone/Socket |