Learn More
Kartell™ Filtration and Vaccum Dessiccators
BIOPHARMA
Product Code. p-8007238
Change view
Click to view available options
Volume (Metric):
2.15 L
4.35 L
9.2 L
Height (Metric):
190 mm
230 mm
300 mm
Height (Metric) Interior:
135 mm
175 mm
225 mm
Diameter (Metric) Outer:
150 mm
200 mm
250 mm
Unit Size:
Each
Product Code. Volume (Metric) Height (Metric) Height (Metric) Interior Diameter (Metric) Outer unitSize
10365821 2.15 L 190 mm 135 mm 150 mm Each
10683272 4.35 L 230 mm 175 mm 200 mm Each
10335921 9.2 L 300 mm 225 mm 250 mm Each
3 options
Product Code. 10365821
Brand: Kartell™ 0055000
Only null left
Add to basket
Add to basket
Can be evacuated to 1bar, with loss not exceeding 0.03mbar in 24 hours. Kartell™ Filtration and Vacuum Dessicators is implosion proof.
  • Transparent polycarbonate cover seals onto a neoprene O-ring set into the flange of the polypropylene base
  • Initial vacuum established with a little grease spread on the O-ring
  • Removable pan for holding a drying agent
  • Includes vacuum release tap/non-return valve assembly
  • Specially grooved stopper in assembly allows air, when required, to re-enter very slowly to avoid disturbing the contents
Specifications
Material Polypropylene, Polycarbonate
Quantity 1 Each
Volume (Metric) 2.15 L
Height (Metric) 190 mm
Height (Metric) Interior 135 mm
Diameter (Metric) Outer 150 mm