Learn More
Labconco™ Filters, Prefilters and Accessories
Accessories for Protector Filtered Glove Boxes, Precise HEPA-Filtered Glove Boxes and XPert Weigh Boxes.
Brand: Labconco™ 5242400
Additional Details : Weight : 2.30000kg
Description
DOP Test Kit for HEPA Filters includes the connections, hoses and test procedure necessary to conduct a HEPA filter leak test on both the inlet and outlet filter. Test should be performed by a qualified technician using calibrated equipment. For use with Labconco Protector Filtered Glove Boxes, Precise HEPA-Filtered Glove Boxes and XPert Weigh Boxes.Specifications
HEPA | |
5 lb. | |
Filter Accessory |
Precise™ HEPA Filtered Glove Box, XPert™ Weigh Box | |
2.3 Kg |