Learn More
Labconco™ Filters, Prefilters and Accessories
Accessories for Protector Filtered Glove Boxes, Precise HEPA-Filtered Glove Boxes and XPert Weigh Boxes.
Brand:  Labconco™ 5242400
Additional Details : Weight : 2.30000kg
 View more versions of this product
Product Code. 17295374
  • 3070.00 NOK / Each
Estimated Shipment: 01-08-2024
to see stock.
Description
Description
DOP Test Kit for HEPA Filters includes the connections, hoses and test procedure necessary to conduct a HEPA filter leak test on both the inlet and outlet filter. Test should be performed by a qualified technician using calibrated equipment. For use with Labconco Protector Filtered Glove Boxes, Precise HEPA-Filtered Glove Boxes and XPert Weigh Boxes.
Specifications
Specifications
HEPA
5 lb.
Filter Accessory
Precise™ HEPA Filtered Glove Box, XPert™ Weigh Box
2.3 Kg