Learn More
Lenz Laborglasintrumente Filter Funnels
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 07195234
Additional Details : Weight : 0.10000kg
Description
- Filter funnels
- Material: borosilicate glass 3.3
Specifications
75 mL | |
130 mm | |
2 |
Borosilicate Glass 3.3 | |
45 mm |