missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Filter Funnels

Made of borosilicate glass. Lenz Laborglasinstrumente™ Filter funnels are ideal for separating solids from liquids via laboratory filtration.

Brand:  Lenz Laborglasinstrumente™ 07195379

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10284291

  • 6693.90 NOK / Each
Estimated Shipment: 10-06-2024
to see stock.
Add to basket

Description

Description

  • Made from borosilicate glass
  • Separates solids from liquids
Specifications

Specifications

4000 mL
400 mm
3
Borosilicate Glass
175 mm
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers