missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Filter Funnel

Robu™ Filter Funnels are designed to be use with a Buchner funnel.

Brand:  Robu™ 21403

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10614302

  • 5100.00 NOK / Each
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

  • Can withstand acids, alkalis, and solvents and even temperatures of up to 540°C
  • Filter sucking
  • Form according to Büchner
Specifications

Specifications

Filter Funnel
4000 mL
Borosilicate Glass
16 to 40 μm
190 mm outer, 180 mm inner
30 mm
206 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers