missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 180 mm

The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.

Brand:  ROBU VitraPOR™ 51800

Additional Details : Weight : 0.48920kg

 View more versions of this product

Product Code. 16843126

  • 2680.00 NOK / Each
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass (3.3)
1 Ea.
13.0 ± 3.0 mm
Non-sterile
0
180 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers