missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 180 mm

The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.

Brand:  ROBU VitraPOR™ 5180F

Additional Details : Weight : 0.48920kg

 View more versions of this product

Product Code. 16893126

  • 3320.00 NOK / Each

Estimated Shipment: 03-01-2023
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass 3.3
1 Ea.
13.0 ± 3.0 mm
Non-sterile
Fine
180 mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers