missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Disc, Surfaces Plane, Ground Cylindrical, 60 mm

The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.

Brand:  ROBU VitraPOR™ 16609

Additional Details : Weight : 0.10450kg

 View more versions of this product

Product Code. 16822866

  • 1285.00 NOK / Pack of 5
Estimated Shipment: 30-05-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
  • The biplane version may be required in specific installation circumstances
Specifications

Specifications

Borosilicate Glass (3.3)
Non-sterile
00
60 mm
00
5/Pk.
4.0 ± 0.2 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers