Learn More
Lenz Laborglasintrumente Filter Crucibles
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 07185130
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10396261
  • 227.90 NOK / Each
Estimated Shipment: 17-06-2024
to see stock.
Add to basket
Description
Description
  • Filter crucibles
  • Material: borosilicate glass 3.3
Specifications
Specifications
Borosilicate Glass (3.3)
30 mL
1