Learn More
ROBU VitraPOR™ Filter Crucible, 50 mL
Description
- For analytical filtration in laboratory
- Suitable glass adapters and rubber sleeves are available
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Quantity | 10/Pk. |
| Capacity (Metric) | 50 mL |
| Porosity | 1 |