Learn More
Hettich Lab Technology™ Filter Cards
Hettich Lab Technology™ Filter Cards are used for collecting cyto centrifuge samples and come in sizes to fit accommodate a variety of chambers and carriers.
Brand:  Hettich Lab Technology™ 1691
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10677682
  • 375.00 NOK / Pack of 100
Estimated Shipment: 28-06-2024
Description
  • For cyto centrifuge systems and cell evaluations
  • Suitable for a variety of cyto chambers and slide carriers
Specifications
Specifications
Filter Card
Slide carrier 1666
2 x 8 mL
Dual channel