Learn More
Hettich Lab Technology™ Filter Cards
Hettich Lab Technology™ Filter Cards are used for collecting cyto centrifuge samples and come in sizes to fit accommodate a variety of chambers and carriers.
Brand: Hettich Lab Technology™ 1691
Additional Details : Weight : 0.01000kg
Description
- For cyto centrifuge systems and cell evaluations
- Suitable for a variety of cyto chambers and slide carriers
Specifications
Filter Card | |
Slide carrier 1666 | |
2 x 8 mL | |
Dual channel |