Learn More
Sartorius Filter Boards
Filter Boards with increased wet strength for the filtration and absorption of liquids.
Marke: Sartorius FT-3-10389-025
Additional Details : Weight : 0.23300kg
Beschreibung
- Made of refined pulp or cotton linters
- Smooth
- Supplied in sheets and discs
Spezifikation
Filter Paper | |
Cotton Linters, Refined Pulp | |
Certified Quality |
Pressure or vacuum filtration | |
Cellulose |