Learn More
Sartorius Filter Boards
Filter Boards with increased wet strength for the filtration and absorption of liquids.
Marke:  Sartorius FT-3-10389-025
Additional Details : Weight : 0.23300kg
Artikelnummer. 15962850
  • 9455.00 NOK / Pack of 1000
Estimated Shipment: 02-07-2024
to see stock.
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Made of refined pulp or cotton linters
  • Smooth
  • Supplied in sheets and discs
Spezifikation
Spezifikation
Filter Paper
Cotton Linters, Refined Pulp
Certified Quality
Pressure or vacuum filtration
Cellulose
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote