Learn More
RETSCH Filter Bags
Brand:  RETSCH 02.186.0004
2356.00 NOK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 11760825
  • 2480.00 NOK / Each
Estimated Shipment: 02-08-2024
Specifications
Filter Bag
For 0.3L TG 200 Drying Container