Learn More
BRAND™ Filling Tubes for BRAND™ Bottletop Dispensers and Burettes
Product Code. 10702202 Shop All Brand Products
Change view
Click to view available options
Length (Metric):
195 to 350 mm
250 mm
250 to 480 mm
500 mm
70 to 140 mm
Sterility:
Non-sterile
For Use With (Equipment):
Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL)
Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (25, 50, 100 mL), Seripettor™ pro (25 mL), Titrette™ (10, 25, 50 mL)
Seripettor™ (All Volumes)
Unit Size:
Each
Pack of 2
Product Code. Length (Metric) Sterility For Use With (Equipment) unitSize
10702202 250 mm Non-sterile Seripettor™ (All Volumes) Pack of 2
15498013 70 to 140 mm Non-sterile Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL) Each
15418023 195 to 350 mm Non-sterile Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL) Each
15428023 250 to 480 mm Non-sterile Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (1, 2, 5, 10 mL), Seripettor™ pro (2, 10 mL) Each
15448023 250 to 480 mm Non-sterile Dispensette™ III, Dispensette™ S, S Organic, and S Trace Analysis (25, 50, 100 mL), Seripettor™ pro (25 mL), Titrette™ (10, 25, 50 mL) Each
10712202 500 mm Non-sterile Seripettor™ (All Volumes) Pack of 2
6 options
Product Code. 10702202
Brand: BRAND™ 704532
Only null left
Add to basket
Add to basket
Replacement filling tubes for BRAND™ bottletop instruments, including Dispensette™ S, Dispensette™ S Organic, and Dispensette™ S Trace Analysis dispensers; seripettor™ and seripettor™ pro dispensers; and Titrette™ bottletop burettes.
  • Telescoping FEP filling tubes adjust to bottle height without cutting
    • Compatible with Dispensette™ S, Dispensette™ S Organic, Dispensette™ S Trace Analysis; seripettor™ pro; and Titrette™
    • Also compatible with Dispensette™ III
  • Standard PP filling tubes can be manually cut to fit various bottle sizes
    • Compatible only with seripettor™
    • Available sterile with O-ring
  • Available in multiple lengths based on dispenser nominal volume
  • Design supports aspiration from the bottom of the bottle while reducing precipitate intake
TRUSTED_SUSTAINABILITY
Specifications
Product Type Filling Tube
Color Clear
Material Polypropylene
For Use With (Equipment) Seripettor™ (All Volumes)
Sterility Non-sterile
Length (Metric) 250 mm