Description
- Anti-static
- Polypropylene has a high resistance to chemicals and temperatures
- The scoops are unbreakable, easy to clean and light-weight
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Green |
| Product Type | Filling Scoop |
| For Use With (Application) | For Industry |
| Width (Metric) | 110 mm |
| Material | Polypropylene |
| Length (Metric) | 250 mm |