Description
- Ready-to-use sterile medium
- Density media optimized for the isolation of lymphocytes from peripheral blood using simple centrifuge-based protocol
- High yield
- Maintains viability and representative distribution of B and T lymphocytes
- Low levels of endotoxin (<0.12 EU/mL)
Specifications
Specifications
| Quantity | 6 x 100 mL |
| Content And Storage | 4–30°C |
| For Use With (Application) | Cell Isolation |
| Endotoxin Level | < 0.12 EU/mL |
| Color | Colorless to Slightly Yellow |
| Product Type | Centrifugation Media |
| Density | 1.078 g/mL |
| Sterility | Sterile |
| Form | Liquid |