missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FICD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11974-1-AP-150UL
This item is not returnable.
View return policy
Description
The function remains known.Specifications
| FICD | |
| Polyclonal | |
| Unconjugated | |
| FICD | |
| AMPylator FICD, FIC domain containing, FIC domain containing protein, FICD, HIP 13, HIP13, Huntingtin yeast partner E, HYPE, UNQ3041 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11153, 231630, 288741 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6AY47, Q8BIX9, Q9BVA6 | |
| Ficd | |
| FICD Fusion Protein Ag2597 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction