missing translation for 'onlineSavingsMsg'
Learn More

Fibulin 5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12188-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16889743

  • 3468.15 NOK / 150µL
Estimated Shipment: 22-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody got 50 kDa or 66 kDa band in western blotting, the 66 kDa corresponding to fully glycosylated protein.

Essential for elastic fiber formation, is involved in the assembly of continuous elastin (ELN) polymer and promotes the interaction of microfibrils and ELN. Stabilizes and organizes elastic fibers in the skin, lung and vasculature. Promotes adhesion of endothelial cells through interaction of integrins and the RGD motif. Vascular ligand for integrin receptors which may play a role in vascular development and remodeling.
Specifications

Specifications

Fibulin 5
Polyclonal
Unconjugated
FBLN5
ARMD3, DANCE, EVEC, FBLN5, FIBL 5, fibulin 5, UP50, Urine p50 protein
Rabbit
Antigen Affinity Chromatography
RUO
10516, 23876, 29158
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UBX5, Q9WVH8, Q9WVH9
Fbln5
Fibulin 5 Fusion Protein Ag2859
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers