missing translation for 'onlineSavingsMsg'
Learn More

Fibromodulin Mouse anti-Human, Clone: 3E9D10, Proteintech

Product Code. 16847075 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16847075 20 μL 20µL
16837075 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16847075

Brand: Proteintech 601081IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fibromodulin is a member of a family of small interstitial proteoglycans, containing a central region composed of leucine-rich repeats with 4 keratan sulfate chains flanked by disulfide-bonded terminal domains. It may participate in the assembly of the extracellular matrix as it interacts with type I and type II collagen fibrils and inhibits fibrillogenesis in vitro. It may also regulate TGF-beta activities by sequestering TGF-beta into the extracellular matrix.

Specifications

Antigen Fibromodulin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3E9D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene FMOD
Gene Accession No. Q06828
Gene Alias fibromodulin, FM, FMOD, KSPG fibromodulin, SLRR2E
Gene Symbols FMOD
Host Species Mouse
Immunogen Fibromodulin Fusion Protein Ag4030
Purification Method Caprylic Acid Precipitation
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2331
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.