Learn More
Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters
Product Code. 15322167
Change view
Click to view available options
Capacity:
10 mL Vacutainer Tube
100 mL Oak Ridge Tube
15 mL Conical Tube
16 mL Oak Ridge Tube
3 mL Vacutainer Tube
30 mL Oak Ridge Tube
Unit Size:
Pack of 2
Product Code. Capacity unitSize
15322167 100 mL Oak Ridge Tube Pack of 2
15332167 30 mL Oak Ridge Tube Pack of 2
15342167 16 mL Oak Ridge Tube Pack of 2
15362167 10 mL Vacutainer Tube Pack of 2
15372167 3 mL Vacutainer Tube Pack of 2
15209653 15 mL Conical Tube Pack of 2
6 options
Product Code. 15322167
Brand: Thermo Scientific™ 0101119
Add versatility to your fixed-angle rotor with Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters, which support various conical tubes.
Add versatility to your fixed-angle rotor with Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters, which support Ridge™, Vacutainer™ and conical tubes.
Adapters hold Oak Ridge™, Vacutainer™ and conical tubes, and vary by capacity.
Ordering Information:
Order Lid O-ring (022062034), O-ring refresher kit (021062038), Lid assembly (099062034) and Fiberlite F14-6x250y Rotor (78500) separately. Contact your Sales Representative for more information.
TRUSTED_SUSTAINABILITY
Tekniske data
Type Rotor Tube Adapter
Capacity 100 mL Oak Ridge Tube
Vessels per Adapter 1
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.