Learn More
Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters
Product Code. 15209653
Change view
Click to view available options
Capacity:
10 mL Vacutainer Tube
100 mL Oak Ridge Tube
15 mL Conical Tube
16 mL Oak Ridge Tube
3 mL Vacutainer Tube
30 mL Oak Ridge Tube
Unit Size:
Pack of 2
Product Code. Capacity unitSize
15209653 15 mL Conical Tube Pack of 2
15332167 30 mL Oak Ridge Tube Pack of 2
15342167 16 mL Oak Ridge Tube Pack of 2
15362167 10 mL Vacutainer Tube Pack of 2
15322167 100 mL Oak Ridge Tube Pack of 2
15372167 3 mL Vacutainer Tube Pack of 2
6 options
Product Code. 15209653
Brand: Thermo Scientific™ 0101410
Add versatility to your fixed-angle rotor with Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters, which support various conical tubes.
Add versatility to your fixed-angle rotor with Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters, which support Ridge™, Vacutainer™ and conical tubes.
Adapters hold Oak Ridge™, Vacutainer™ and conical tubes, and vary by capacity.
Ordering Information:
Order Lid O-ring (022062034), O-ring refresher kit (021062038), Lid assembly (099062034) and Fiberlite F14-6x250y Rotor (78500) separately. Contact your Sales Representative for more information.
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Tube Adapter
Capacity 15 mL Conical Tube
Vessels per Adapter 5