Learn More
Brady™ Fiberglass-Reinforced Nylon Miniature Locking System for Circuit Breakers
A safe and effective method for locking out European Circuit Breakers.
Brand: Brady™ 090848
Additional Details : Weight : 0.05580kg
Description
- Requires no tools to install
- Lockout easily installs with the aid of a push button
- Tie-bar style lockout has a thumb wheel for quick application
Specifications
Miniature Locking System for Circuit Breakers | |
Fiberglass-reinforced Nylon | |
24.13 mm | |
Pin In | |
24.13 mm |
Red/Black | |
Miniature Circuit Breaker Lockouts - Pin-In Standard | |
13.72 x 24.13 x 41.7 mm | |
6 Pk. |