Learn More
Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Product Code. 15810530 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
25 (5 x 5 Grid)
49 (7 x 7 Grid)
64 (8 x 8 Grid)
81 (9 x 9 Grid)
Quantity:
12 Pack
Each
Unit Size:
Each
Pack of 12
Product Code. Holds Quantity unitSize
15810530 64 (8 x 8 Grid) 12 Pack Pack of 12
12744388 49 (7 x 7 Grid) 12 Pack Each
11849490 81 (9 x 9 Grid) 12 Pack Pack of 12
15907976 25 (5 x 5 Grid) Each Each
15821152 49 (7 x 7 Grid) Each Each
11329341 81 (9 x 9 Grid) Each Each
6 options
Product Code. 15810530
Brand: Thermo Scientific™ 5960
Only null left
Add to basket
Add to basket
Organize vial storage within cryo boxes with these fiberboard box dividers.
Organize vial storage with Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers.
  • Use with fiberboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
TRUSTED_SUSTAINABILITY
Specifications
Quantity 12 Pack
Holds 64 (8 x 8 Grid)