Learn More
Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Product Code. 11849490 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
25 (5 x 5 Grid)
49 (7 x 7 Grid)
64 (8 x 8 Grid)
81 (9 x 9 Grid)
Quantity:
12 Pack
Each
Unit Size:
Each
Pack of 12
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Quantity Unit Size
11849490 81 (9 x 9 Grid) 12 Pack Pack of 12
12744388 49 (7 x 7 Grid) 12 Pack Each
15810530 64 (8 x 8 Grid) 12 Pack Pack of 12
15907976 25 (5 x 5 Grid) Each Each
15821152 49 (7 x 7 Grid) Each Each
11329341 81 (9 x 9 Grid) Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11849490 Supplier Thermo Scientific™ Supplier No. AVL6212
  • ONLINE EXCLUSIVE PRICE
    395.00 NOK
    386.89 NOK / Pack of 12
    Save 8.11 NOK
    2% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
Organize vial storage within cryo boxes with these fiberboard box dividers.
Organize vial storage with Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers.
  • Use with fiberboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
TRUSTED_SUSTAINABILITY
Specifications
Quantity 12 Pack
Holds 81 (9 x 9 Grid)