Learn More
SCHOTT AG Lighting and Imaging™ Fiber Optic Flexible Light Guide
Product Code. 15773012 Shop All Schott Products
Click to view available options
Includes:
Double Lightguide
Single Lightguide
Triple Lightguide
Diameter (Metric) Inner:
4.5 mm
5 mm
8 mm
9 mm
Length (Metric):
1 m
1.6 m
Unit Size:
1 unit
Each
Product Code. 15773012
Brand: SCHOTT AG Lighting and Imaging™ 155206
Fiber Optic Flexible Light Guide in a black metal-PVC jacketing.
SCHOTT Flexible Light Guides are highly bendable. In contrast to goosenecks, they allow a higher flexibility and need to be held.
  • Light guide with eco-friendly Puravis fiber optics.
  • Available in different lengths and diameters.
  • Available with one, two or three branches.
  • High white light transmission thanks to the use of SCHOTT PURAVIS™ fibers.
  • High flexibility for different lighting, as well as remote illumination.
Specifications
Type Flexible Fiber Optic Light Guide
Diameter (Metric) Inner 4.5 mm
For Use With (Equipment) Cold Light Source KL1600LED, KL2500LED or KL1500HAL
Includes Triple Lightguide
Length (English) 3.3 ft.
Length (Metric) 1 m
Material Eco-friendly Puravis Glass Optical Fibers
Quantity 1 Ea.