missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHL1 Rabbit anti-Human, Mouse, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10991-1-AP-150UL
This item is not returnable.
View return policy
Description
This is a rabbit polyclonal antibody raised against full length of human FHL1.
This gene encodes a member of the four-and-a-half-LIM-only protein family. Family members contain two highly conserved, tandemly arranged, zinc finger domains with four highly conserved cysteines binding a zinc atom in each zinc finger. Expression of these family members occurs in a cell- and tissue-specific mode and these proteins are involved in many cellular processes. Mutations in this gene have been found in patients with Emery-Dreifuss muscular dystrophy. Multiple alternately spliced transcript variants which encode different protein isoforms have been described.Specifications
| FHL1 | |
| Polyclonal | |
| Unconjugated | |
| FHL1 | |
| FHL 1, FHL1, FHL1B, FLH1A, four and a half LIM domains 1, KYO T, Skeletal muscle LIM protein 1, SLIM, SLIM 1, SLIM1, XMPMA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14199, 2273, 387528, 399646 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P97447, Q13642 | |
| FHL1, fhl1a, fhl1b | |
| FHL1 Fusion Protein Ag1448 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction