missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR5/FGFRL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
1975.00 NOK - 2985.00 NOK
Specifications
| Antigen | FGFR5/FGFRL1 |
|---|---|
| Dilution | ELISA 1:1000-1:2000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18653652
|
Novus Biologicals
NBP2-98227-100ul |
100 μg |
2985.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654702
|
Novus Biologicals
NBP2-98227-50ul |
50 μg |
1975.00 NOK
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGFR5/FGFRL1 Polyclonal antibody specifically detects FGFR5/FGFRL1 in Mouse samples. It is validated for ELISASpecifications
| FGFR5/FGFRL1 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis | |
| 0.2 um filtered solution in PBS | |
| 53834 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| FGF homologous factor receptor, FGF receptor-like protein 1, FGFR5, FGFR-5, FGFR-like protein, FHFR, Fibroblast growth factor receptor 5, fibroblast growth factor receptor-like 1 | |
| Produced in rabbits immunized with purified, recombinant Mouse FGFR5/FGFRL1 (Accession#: NP_473412.1; Met 1-Pro 374) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title