Learn More
Invitrogen™ Human FGFR3 [V555M], GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This FGFR3 V555M recombinant human protein (397-end) was expressed in insect cells. FGFR3 V555M recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Cytoplasmic, Mutant The gene accession number for FGFR3 [V555M] is NP_000133.1.
Specifications
Specifications
| Accession Number | P22607 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris HCl with 25% glycerol, 0.1mM PMSF, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA and no preservative; pH 7.5 |
| Gene ID (Entrez) | 2261 |
| Molecular Weight (g/mol) | 71.8 kDa |
| Name | Human FGFR3 [V555M], GST Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.