missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FGFR3, His Tag Recombinant Protein

FGFR3 is a member of the FGFR family of receptor tyrosine kinases.

Brand:  Invitrogen™ PV4107

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 10352163

  • 69400.00 NOK / 1mg
Estimated Shipment: 27-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The FGFR family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy.

Specifications

Specifications

NP_000133
50mM Tris, pH 7.5, 100mM NaCl, 0.2mM EDTA, 0.05% Triton X-100, 5mM DTT, 50% glycerol
1mg
RUO - research use only
FGFR3, CEK2, JTK4
Human
His Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
49.8kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
FGF Receptor Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers