missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF basic/FGF2/bFGF Antibody (HuGAL-F2) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28540JF669
This item is not returnable.
View return policy
Description
FGF basic/FGF2/bFGF Monoclonal antibody specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| FGF basic/FGF2/bFGF | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2 | |
| FGF2 | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Cancer | |
| 2247 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| HuGAL-F2 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction