Få mere at vide
Chemsplash FFP3 Fold Flat Mask with Valve
Beskrivelse
Chemsplash 3675 P3 Fold Flat Mask
- High Filtration Capacity
- Respirator with Red Color Exhalation Valve
- Exhalation Valve Reduces Hot Air Buildup
- Woven Elasticated Adjustable Head Straps
- Adjustable Nose Clip with Soft Inner Nose Foam
- Folded to Pocket Size and Packed Individually for Hygiene
- Protects Against Harmful and Toxic Solid and Liquid Particles
- No Vision Impairment and Problem-Free Speech
- Latex and Graphene Free
Description: The Chemsplash 3675 Fold Flat Mask is a P3 valved fold-flat respirator, color-coded red for easy identification. This mask offers a high filtration capacity and features woven elasticated adjustable toggles for a comfortable fit. It includes an adjustable nose clip with soft inner foam to ensure an excellent seal, minimizing air leakage. The mask is folded to pocket size and packed individually for hygiene. It effectively protects against harmful and toxic solid and liquid particles, ensuring no vision impairment or speech difficulty. Additionally, it is latex and graphene-free for safer use.
Tekniske data
Tekniske data
| Aerosol Type | R - Somewhat Resistant to Oil |
| Color | White |
| Design Type | Flat-fold |
| Disposable | Single-use |
| Filter Type | FFP3 |
| Material | Polypropylene |
| Product Type | General Purpose Particulate Respirator |
| Size | One Size |
| With Exhalation Valve | Yes |
| Latex Free | Yes |
| Vis mere |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.