Learn More
Chemsplash FFP2 Cup Mask with Valve
Artikelnummer. 30058359
Ansicht ändern
Click to view available options
Aerosol Type:
N - Not Resistant to Oil
Packungsgröße:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Aerosol Type Packungsgröße
30058359 N - Not Resistant to Oil Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30058359 Lieferant Chemsplash Lieferanten-Nr. 3672
  • ONLINE EXCLUSIVE PRICE
    1517.63 NOK
    1283.23 NOK / Pack of 200
    Save 234.40 NOK
    15% Off
Estimated Shipment: 15-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
The FFP2 valved cup mask has a high filtering efficiency. It has woven elasticated adjustable head straps.
Chemsplash 3672 FFP2 Valved Cup Mask
  • High Filtration Capacity
  • Respirator with Yellow Color Exhalation Valve
  • Exhalation Valve Reduces Hot Air Buildup
  • Woven Elasticated Adjustable Head Straps
  • Adjustable Nose Clip with Soft Inner Nose Foam
  • Protects Against Low-Toxicity Solid and Liquid Particles
  • No Vision Impairment and Problem-Free Speech
  • Latex and Graphene Free
Description: The Chemsplash 3672 is an FFP2 valved cup mask, colour-coded yellow for ease of use. The fabric material of the mask has a high filtering efficiency. It features woven elasticated adjustable head straps and a soft padded band that rests inside, adjustable for an excellent fit on the nose. This mask provides reliable protection against low-toxicity solid and liquid particles while ensuring no impairment to vision and problem-free speech. It is latex and graphene-free for enhanced safety and comfort.
Spezifikation
Aerosol Type N - Not Resistant to Oil
Color White
Design Type Cup/Molded
Disposable Single-use
Filter Type FFP2
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve Yes
Latex Free Yes
Sterility Non-sterile
Mehr anzeigen Weniger anzeigen