missing translation for 'onlineSavingsMsg'
Learn More

FFAR2/GPR43 Antibody - BSA Free, Novus Biologicals™

Product Code. 18515737 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
Unit Size:
50µL
Product Code. Quantity unitSize
18515737 0.05 mL 50µL
1 options
This item is not returnable. View return policy

Product Code. 18515737

Brand: Novus Biologicals NLS6599

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FFAR2/GPR43 Polyclonal antibody specifically detects FFAR2/GPR43 in Human, Monkey, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FFAR2/GPR43
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 5 - 10 μg/mL
Formulation PBS
Gene Alias free fatty acid activated receptor 2, free fatty acid receptor 2, G protein-coupled receptor 43, GPR43FFA2R, G-protein coupled receptor 43
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from 3rd extracellular domain of human FFAR2/GPR43.
Purification Method Immunogen affinity purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2867
Target Species Human, Monkey, Primate
Content And Storage Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.